Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Pinnacle Portfolio Clarifier™ 1, 2

Catalog No. 22220107 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Clarifier 1
Clarifier 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-220-107 Clarifier 1
22-220-108 Clarifier 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-220-107
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unique acetic acid reagents, developed to supplement our family of Hematoxylins

  • Designed as a system
  • Removes background stain caused by excessive use of tissue adhesive Increases cell transparency Selectively removes hematoxylin staining caused by excess adhesive without affecting nuclear staining
  • Use of Clarifier will improve contrast between the nuclei and cytoplasm
  • Acid alcohol requires quick dips while Clarifier dictates a set time (30-60 seconds for Clarifier 1 and 20-30 seconds for Clarifier 2)
  • Each component has added value and benefits that reduces cost per slide
  • Unique chemical formulae promotes long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity

Specifications

Color Clear
Format Liquid
For Use With (Application) Developed to supplement our family of hematoxylins
Quantity 4 x 1 gal.
Product Type Clarifier 1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.