Learn More
TARGETMOL CHEMICALS INC Pinobanksin 3-acetate 5MG
SDP
Catalog No. 502876541
Encompass_Preferred
Also available in 1 mg, 10 mg and bulk. Please contact Fisher for quotes. Pinobanksin 3-acetate (3-O-Acetylpinobanksin), a flavonoid derivative, is a strong antioxidant found in sunflowers and honey that inhibits LDL peroxidation. Purity 99.79%
Catalog No. 50-287-6541 Supplier TARGETMOL CHEMICALS INC Supplier No. TN29675MG
Also available in 1 mg, 10 mg and bulk. Please contact Fisher for quotes. Pinobanksin 3-acetate (3-O-Acetylpinobanksin), a flavonoid derivative, is a strong antioxidant found in sunflowers and honey that inhibits LDL peroxidation. Purity 99.79%