Learn More
TARGETMOL CHEMICALS INC PINOCEMBRIN 25MG
SDP
Catalog No. 502580444
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pinocembrin (57-Dihydroxyflavanone) is a flavanone a type of flavonoid. It is an antioxidant found in damiana honey fingerroot and propolis. purity: 98%
Catalog No. 50-258-0444 Supplier TARGETMOL CHEMICALS INC Supplier No. T299225MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pinocembrin (57-Dihydroxyflavanone) is a flavanone a type of flavonoid. It is an antioxidant found in damiana honey fingerroot and propolis. purity: 98%