Learn More
Cayman Chemical PInocembrIn 50mg
SDP
Supplier:  Cayman Chemical 2985250 MG
Encompass_Preferred
A flavonoid with diverse biological activities; induces apoptosis in, and inhibits the migration of, SKOV3 ovarian cancer cells at 200 µM; reduces lesion volume, as well as brain microglial activation and production of IL-1β, IL-6, and TNF-α in a mouse model of collagenase-induced ICH at 5 mg/kg; prevents increases in plasma and kidney MDA levels, glomeruli lobulation, mesangial expansion, and tubule vacuolization and occlusion, and it decreases hepatic cholesterol, triglyceride, and LDL levels in a rat model of diabetic nephropathy; reduces pulmonary edema, as well as neutrophil, lymphocyte, and macrophage infiltration in a mouse model of LPS-induced lung injury at 20 and 50 mg/kg
Catalog No. 50-298-8863