Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinocembrin-7-O-β-D-glucopyranoside | 75829-43-5 | 10 MM 1 ML
SDP

Catalog No. 5000418785
Encompass_Preferred

Pinocembrin-7-O-β-D-glucopyranoside is a flavanone that enhances lipid peroxidation. It is isolated from the dried whole plants of Viscum articulatum and is for research use only.

  • High purity (99.87%)
  • Molecular formula C21H22O9
  • Molecular weight 418.39
  • White to off-white solid appearance
  • Enhances lipid peroxidation
  • Inhibits LPS-induced nitric oxide production

Catalog No. 50-004-18785 Supplier Medchemexpress LLC Supplier No. HYN661610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinocembrin-7-O-β-D-glucopyranoside is a flavanone that enhances lipid peroxidation. It is isolated from the dried whole plants of Viscum articulatum and is for research use only.

  • High purity (99.87%)
  • Molecular formula C21H22O9
  • Molecular weight 418.39
  • White to off-white solid appearance
  • Enhances lipid peroxidation
  • Inhibits LPS-induced nitric oxide production

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.