Learn More
Medchemexpress LLC Pinocembrin-7-O-β-D-glucopyranoside | 75829-43-5 | 5 MG
SDP
Catalog No. 5000419364
Encompass_Preferred
Pinocembrin-7-O-β-D-glucopyranoside is a flavanone isolated from the dried whole plants of Viscum articulatum. It enhances lipid peroxidation and exhibits anti-inflammatory activity.
  • Flavanone compound
  • Enhances lipid peroxidation
  • Isolated from dried whole plants of Viscum articulatum
  • Exhibits anti-inflammatory activity against mouse RAW264.7 cells
  • Inhibits LPS-induced nitric oxide production
  • For research use only
Catalog No. 50-004-19364 Supplier Medchemexpress LLC Supplier No. HYN66165MG
Pinocembrin-7-O-β-D-glucopyranoside is a flavanone isolated from the dried whole plants of Viscum articulatum. It enhances lipid peroxidation and exhibits anti-inflammatory activity.
  • Flavanone compound
  • Enhances lipid peroxidation
  • Isolated from dried whole plants of Viscum articulatum
  • Exhibits anti-inflammatory activity against mouse RAW264.7 cells
  • Inhibits LPS-induced nitric oxide production
  • For research use only