Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pinocembrin, 95% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111004050 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4050 Supplier MilliporeSigma Supelco Supplier No. P523950MG
Add to Cart
Add to Cart

Anti-inflammatory.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Solid
Linear Formula C15H12O4
MDL Number MFCD06858345
Melting Point 194°C to 195°C
Type Analytical Standard
Quantity 50 mg
Formula Weight 256.25
Percent Purity 95% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.