Learn More
TARGETMOL CHEMICALS INC Pinocembrin chalcone 5MG
SDP
Catalog No. 502879715
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Pinocembrin chalcone is an inhibitor of tyrosinase with antimutagenic effects. Pinocembrin chalcone can be used in studies about the prevention of gastric ulcers. Purity 97.82%
Catalog No. 50-287-9715 Supplier TARGETMOL CHEMICALS INC Supplier No. TN20785MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Pinocembrin chalcone is an inhibitor of tyrosinase with antimutagenic effects. Pinocembrin chalcone can be used in studies about the prevention of gastric ulcers. Purity 97.82%