Learn More
Cayman Chemical Pinolenic Acid ethyl ester, Purity: ≥98%, CAS: 493015-74-0, 1 mg
SDP
Catalog No. 502889893
Encompass_Preferred
A polyunsaturated fatty acid ethyl ester; decreases blood glucose levels in an oral glucose tolerance test in mice at 100 mg/kg
Catalog No. 50-288-9893 Supplier Cayman Chemical Supplier No. 100086581 MG
A polyunsaturated fatty acid ethyl ester; decreases blood glucose levels in an oral glucose tolerance test in mice at 100 mg/kg