Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinolenic acid ethyl ester | 493015-74-0 | 98.0% | 306.48 g/mol | C20H34O2 | 25MG
SDP

Catalog No. 5000239048
Encompass_Preferred

Pinolenic acid ethyl ester is a polyunsaturated fatty acid ethyl ester supplied as a high-purity research reagent for biochemical and lipid metabolism studies. It is suitable as an analytical standard for assay development, structure-activity investigations, and formulation research. Follow the safety data sheet for handling and storage instructions.

  • High purity: 98.0%.
  • Molecular formula: C20H34O2.
  • Molecular weight: 306.48 g/mol.
  • CAS registry number: 493015-74-0.
  • Offered in small research pack sizes for analytical use.
  • Intended for laboratory research use only; not for human consumption.

Catalog No. 50-002-39048 Supplier Medchemexpress LLC Supplier No. HY12418725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pinolenic acid ethyl ester is a polyunsaturated fatty acid ethyl ester supplied as a high-purity research reagent for biochemical and lipid metabolism studies. It is suitable as an analytical standard for assay development, structure-activity investigations, and formulation research. Follow the safety data sheet for handling and storage instructions.

  • High purity: 98.0%.
  • Molecular formula: C20H34O2.
  • Molecular weight: 306.48 g/mol.
  • CAS registry number: 493015-74-0.
  • Offered in small research pack sizes for analytical use.
  • Intended for laboratory research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.