Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinolenic acid ethyl ester | 493015-74-0 | MFCD12912314 | ≥98.0% | 306.48 g·mol⁻1 | C20H34O2 | 5MG
SDP

Catalog No. 5000236779
Encompass_Preferred

Pinolenic acid ethyl ester is the ethyl ester derivative of pinolenic acid, a polyunsaturated fatty acid found in pine seed oils. It is supplied as a research reagent for biochemical and lipid metabolism studies where a more lipophilic form of the free acid is required.

  • Ethyl ester of pinolenic acid (CAS 493015-74-0).
  • Purity ≥98.0%.
  • Molecular formula C20H34O2.
  • Molecular weight 306.48 g·mol⁻1.
  • Appearance colorless to light yellow liquid; density ≈0.893 g/cm³.
  • Storage store at 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Typical pack sizes 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.

Catalog No. 50-002-36779 Supplier Medchemexpress LLC Supplier No. HY1241875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinolenic acid ethyl ester is the ethyl ester derivative of pinolenic acid, a polyunsaturated fatty acid found in pine seed oils. It is supplied as a research reagent for biochemical and lipid metabolism studies where a more lipophilic form of the free acid is required.

  • Ethyl ester of pinolenic acid (CAS 493015-74-0).
  • Purity ≥98.0%.
  • Molecular formula C20H34O2.
  • Molecular weight 306.48 g·mol⁻1.
  • Appearance colorless to light yellow liquid; density ≈0.893 g/cm³.
  • Storage store at 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Typical pack sizes 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.