Learn More
Cayman Chemical Pinolenic Acid ethyl ester
SDP
Catalog No. 502887737
Encompass_Preferred
A polyunsaturated fatty acid ethyl ester; decreases blood glucose levels in an oral glucose tolerance test in mice at 100 mg/kg
Catalog No. 50-288-7737 Supplier Cayman Chemical Supplier No. 1000865810 MG
A polyunsaturated fatty acid ethyl ester; decreases blood glucose levels in an oral glucose tolerance test in mice at 100 mg/kg