Learn More
Cayman Chemical Pinolenic Acid Fatty Acids and
SDP
Catalog No. 502889026
Encompass_Preferred
Pinolenic acid is a PUFA found in Korean pine (Pinus orientalis) and maritime pine (Pinus pinaster) seed oils. Both oils have been found to have lipid-lowering properties. A diet containing maritime pine seed oil (MPSO) lowered HDL and ApoA1 levels in transgenic mice expressing human ApoA1. MPSO was found to diminish cholesterol efflux in vitro.{14273} Korean pine seed oil supplements may help in obesity by reduction of appetite. People taking this oil had an increase in the satiety hormones CCK and GLP-1 and a reduced desire to EAt.{14280} The activity of the oil is attributed to pinolenic acid. Pinolenic acid is not converted to arachidonic acid metabolically and can reduce arachidonic acid levels in the phosphatidylinositol fraction of Hep-G2 cells from 15.9% to 7.0%.{8724}
Catalog No. 50-288-9026 Supplier Cayman Chemical Supplier No. 100086541 MG
Add to Cart
Add to Cart
Pinolenic acid is a PUFA found in Korean pine (Pinus orientalis) and maritime pine (Pinus pinaster) seed oils. Both oils have been found to have lipid-lowering properties. A diet containing maritime pine seed oil (MPSO) lowered HDL and ApoA1 levels in transgenic mice expressing human ApoA1. MPSO was found to diminish cholesterol efflux in vitro.{14273} Korean pine seed oil supplements may help in obesity by reduction of appetite. People taking this oil had an increase in the satiety hormones CCK and GLP-1 and a reduced desire to EAt.{14280} The activity of the oil is attributed to pinolenic acid. Pinolenic acid is not converted to arachidonic acid metabolically and can reduce arachidonic acid levels in the phosphatidylinositol fraction of Hep-G2 cells from 15.9% to 7.0%.{8724}