Learn More
Cayman Chemical Pinolenic Acid methyl ester
SDP
Catalog No. 502888991
Encompass_Preferred
Pinolenic acid methyl ester is a neutral, more lipophilic form of the free acid.
Catalog No. 50-288-8991 Supplier Cayman Chemical Supplier No. 100086561 MG
Add to Cart
Add to Cart
Pinolenic acid methyl ester is a neutral, more lipophilic form of the free acid.