Learn More
Medchemexpress LLC HY-15593 10mg Medchemexpress, Pinometostat CAS:1380288-87-8 Purity:>98%
SDP
Catalog No. 502029144
Encompass_Preferred
Medchemexpress, HY-15593 10mg Pinometostat CAS:1380288-87-8 Pinometostat (EPZ-5676) is a potent DOT1L histone methyltransferase inhibitor with a Ki of 80 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9144 Supplier Medchemexpress LLC Supplier No. HY1559310MG
Medchemexpress, HY-15593 10mg Pinometostat CAS:1380288-87-8 Pinometostat (EPZ-5676) is a potent DOT1L histone methyltransferase inhibitor with a Ki of 80 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.