Learn More
Cayman Chemical (−)-Pinoresinol, Purity: ≥95%, CAS: 81446-29-9, 10 mg
SDP
Catalog No. 502909766
Encompass_Preferred
A lignan with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (EC50s = 0.2 and 0.055 mg/ml, respectively); selectively inhibits 5-LO and 15-LO over COX-2 (IC50s = 13.8, 3.5, and >100 µM, respectively); inhibits LPS-induced NO production in BV-2 microglial cells (IC50 = 34.25 µM)
Catalog No. 50-290-9766 Supplier Cayman Chemical Supplier No. 3685010 MG
A lignan with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (EC50s = 0.2 and 0.055 mg/ml, respectively); selectively inhibits 5-LO and 15-LO over COX-2 (IC50s = 13.8, 3.5, and >100 µM, respectively); inhibits LPS-induced NO production in BV-2 microglial cells (IC50 = 34.25 µM)