Learn More
Medchemexpress LLC Pinoresinol | 487-36-5 | 5 MG
SDP
Catalog No. 5000418010
Encompass_Preferred
Pinoresinol is a lignol of plant origin that serves for defense in caterpillars. It significantly sensitizes cancer cells against TNF-related apoptosis-inducing ligand (TRAIL)-induced apoptosis. In vitro studies showed that Pinoresinol facilitated death-inducing signaling complex formation to trigger a caspase-8-dependent apoptotic cascade activation in TRAIL-resistant glioblastoma cells.
  • Lignol of plant origin
  • Serves for defense in caterpillars
  • Sensitizes cancer cells against TRAIL-induced apoptosis
  • Facilitates death-inducing signaling complex formation
  • Triggers caspase-8-dependent apoptotic cascade activation
Catalog No. 50-004-18010 Supplier Medchemexpress LLC Supplier No. HYN62535MG
Pinoresinol is a lignol of plant origin that serves for defense in caterpillars. It significantly sensitizes cancer cells against TNF-related apoptosis-inducing ligand (TRAIL)-induced apoptosis. In vitro studies showed that Pinoresinol facilitated death-inducing signaling complex formation to trigger a caspase-8-dependent apoptotic cascade activation in TRAIL-resistant glioblastoma cells.
  • Lignol of plant origin
  • Serves for defense in caterpillars
  • Sensitizes cancer cells against TRAIL-induced apoptosis
  • Facilitates death-inducing signaling complex formation
  • Triggers caspase-8-dependent apoptotic cascade activation