Learn More
Medchemexpress LLC HY-N2180 5mg Medchemexpress, Pinoresinol dimethyl ether CAS:29106-36-3 Purity:>98%
SDP
Catalog No. 501970372
Encompass_Preferred
Medchemexpress, HY-N2180 5mg Pinoresinol dimethyl ether CAS:29106-36-3 Pinoresinol dimethyl ether ((+)-Eudesmin) is a non-phenolic furofuran lignan isolated from the stem bark of Magnolia kobus with neuritogenic activity. Pinoresinol dimethyl ether ((+)-Eudesmin) can induce neuritis outgrowth from PC12 cells by stimulating up-stream MAPK, PKC and PKA pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0372 Supplier Medchemexpress LLC Supplier No. HYN21805MG
Medchemexpress, HY-N2180 5mg Pinoresinol dimethyl ether CAS:29106-36-3 Pinoresinol dimethyl ether ((+)-Eudesmin) is a non-phenolic furofuran lignan isolated from the stem bark of Magnolia kobus with neuritogenic activity. Pinoresinol dimethyl ether ((+)-Eudesmin) can induce neuritis outgrowth from PC12 cells by stimulating up-stream MAPK, PKC and PKA pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.