Learn More
Medchemexpress LLC Pinoresinol dimethyl ether (+)-eudesmin | 29106-36-3 | MFCD09992396 | 99.9% | 386.44 g/mol | C22H26O6 | 10 MG
SDP
Catalog No. 5000296047
Encompass_Preferred
Pinoresinol dimethyl ether ((+)-Eudesmin) is a furofuran lignan provided as a research-grade chemical standard. It is a solid, white to off-white compound used as an analytical standard and in vitro tool compound reported to modulate S6K1 signaling and to exhibit anti-inflammatory and antibacterial activities.
  • High purity (>99.9%) suitable for analytical and in vitro studies.
  • Solid white to off-white powder for easy handling and weighing.
  • Highly soluble in DMSO (≈100 mg/mL); ultrasonic assistance recommended.
  • Store protected from light at 4°C; solutions stable up to 6 months at -80°C.
  • Available in small pack sizes optimized for research use.
Catalog No. 50-002-96047 Supplier Medchemexpress LLC Supplier No. HYN218010MG
Pinoresinol dimethyl ether ((+)-Eudesmin) is a furofuran lignan provided as a research-grade chemical standard. It is a solid, white to off-white compound used as an analytical standard and in vitro tool compound reported to modulate S6K1 signaling and to exhibit anti-inflammatory and antibacterial activities.
  • High purity (>99.9%) suitable for analytical and in vitro studies.
  • Solid white to off-white powder for easy handling and weighing.
  • Highly soluble in DMSO (≈100 mg/mL); ultrasonic assistance recommended.
  • Store protected from light at 4°C; solutions stable up to 6 months at -80°C.
  • Available in small pack sizes optimized for research use.