Learn More
Cayman Chemical Pinostilbene, Purity: ≥98%, CAS: 42438-89-1, 10 mg
SDP
Catalog No. 502955365
Encompass_Preferred
A stable, monomethoxylated resveratrol derivative that at 0.1-10 μM reduces neurotoxicity in SH-SY5Y cells induced by the parkinsonian mimetic 6-OHDA and scavenges DPPH radicals (IC50 = 47.1 μM)
Catalog No. 50-295-5365 Supplier Cayman Chemical Supplier No. 1404410 MG
A stable, monomethoxylated resveratrol derivative that at 0.1-10 μM reduces neurotoxicity in SH-SY5Y cells induced by the parkinsonian mimetic 6-OHDA and scavenges DPPH radicals (IC50 = 47.1 μM)