Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pinostilbene 97.0+%, TCI America™
SDP

Catalog No. P1927100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1927100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1927100MG Supplier TCI America Supplier No. P1927100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42438-89-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD12407153,MFCD20527315
InChI Key KUWZXOMQXYWKBS-UHFFFAOYSA-N
Synonym 3,4′C-Dihydroxy-5-methoxy-trans-stilbene
PubChem CID 5473050
ChEBI CHEBI:63672
IUPAC Name 3-[2-(4-hydroxyphenyl)ethenyl]-5-methoxyphenol
SMILES COC1=CC(C=CC2=CC=C(O)C=C2)=CC(O)=C1

Specifications

Melting Point 119°C
Color White
Quantity 100 mg
Formula Weight 242.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pinostilbene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.