Learn More
Medchemexpress LLC Pinostilbene (trans-Pinostilbene) | 42438-89-1 | 5 MG
SDP
Catalog No. 5000418346
Encompass_Preferred
Pinostilbene (trans-Pinostilbene) is a major metabolite of Pterostilbene that has shown inhibitory effects on colon cancer cells. This compound is intended for research use only.
  • Exhibits inhibitory effects on colon cancer cells.
  • Does not significantly inhibit the growth of normal colon cells.
  • Causes a significant and dose-dependent increase in the percentage of cells in S phase in colon cancer cells.
  • Modulates expression of key signaling proteins related to cell proliferation and apoptosis.
  • Displays protective effects against 6-hydroxydopamine-induced neurotoxicity.
Catalog No. 50-004-18346 Supplier Medchemexpress LLC Supplier No. HYN30595MG
Pinostilbene (trans-Pinostilbene) is a major metabolite of Pterostilbene that has shown inhibitory effects on colon cancer cells. This compound is intended for research use only.
  • Exhibits inhibitory effects on colon cancer cells.
  • Does not significantly inhibit the growth of normal colon cells.
  • Causes a significant and dose-dependent increase in the percentage of cells in S phase in colon cancer cells.
  • Modulates expression of key signaling proteins related to cell proliferation and apoptosis.
  • Displays protective effects against 6-hydroxydopamine-induced neurotoxicity.