Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinostilbene (trans-pinostilbene) | 42438-89-1 | MFCD20527315 | 100.0% | 242.27 g/mol | C15H14O3 | 10 MG
SDP

Catalog No. 5000289603
Encompass_Preferred

Pinostilbene (trans-pinostilbene) is a metabolite of pterostilbene supplied for research use. It has reported inhibitory effects on colon cancer cells and is commonly used in in vitro cancer assays and metabolite studies. The compound is a high-purity, off-white to light yellow solid suitable for biochemical research.

  • Metabolite of pterostilbene with reported antiproliferative activity in colon cancer cells.
  • High purity: 99.96%.
  • Molecular formula C15H14O3 and molecular weight 242.27 g/mol.
  • Appearance: off-white to light yellow solid.
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Provided as a 10 mg research-grade package.

Catalog No. 50-002-89603 Supplier Medchemexpress LLC Supplier No. HYN305910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinostilbene (trans-pinostilbene) is a metabolite of pterostilbene supplied for research use. It has reported inhibitory effects on colon cancer cells and is commonly used in in vitro cancer assays and metabolite studies. The compound is a high-purity, off-white to light yellow solid suitable for biochemical research.

  • Metabolite of pterostilbene with reported antiproliferative activity in colon cancer cells.
  • High purity: 99.96%.
  • Molecular formula C15H14O3 and molecular weight 242.27 g/mol.
  • Appearance: off-white to light yellow solid.
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Provided as a 10 mg research-grade package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.