Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinostrobin chalcone | 18956-15-5 | 1 MG
SDP

Catalog No. 5000421208
Encompass_Preferred

Pinostrobin chalcone is a potent natural cytotoxic compound, demonstrating efficacy against MDA-MB-231 and HT-29 colon cancer cell lines with IC50 values of 20.42±2.23 and 22.51±0.42 μg/mL, respectively. This product is intended for research use only.

  • Purity: 99.94%
  • Molecular weight: 270.28
  • Formula: C16H14O4
  • Appearance: Solid
  • Color: Light yellow to orange
  • Structure classification: Flavonoids, chalcones, phenols, polyphenols
  • Initial source: Plants, Leguminosae, Cajanus cajan (L.) Millsp.
  • Exhibits anti-inflammatory activity against mouse J774.A1 cells
  • Shows anti-inflammatory activity against mouse RAW264.7 cells
  • Tested for antiproliferative activity against MCF-7 human breast cancer cells

Catalog No. 50-004-21208 Supplier Medchemexpress LLC Supplier No. HYN30571MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinostrobin chalcone is a potent natural cytotoxic compound, demonstrating efficacy against MDA-MB-231 and HT-29 colon cancer cell lines with IC50 values of 20.42±2.23 and 22.51±0.42 μg/mL, respectively. This product is intended for research use only.

  • Purity: 99.94%
  • Molecular weight: 270.28
  • Formula: C16H14O4
  • Appearance: Solid
  • Color: Light yellow to orange
  • Structure classification: Flavonoids, chalcones, phenols, polyphenols
  • Initial source: Plants, Leguminosae, Cajanus cajan (L.) Millsp.
  • Exhibits anti-inflammatory activity against mouse J774.A1 cells
  • Shows anti-inflammatory activity against mouse RAW264.7 cells
  • Tested for antiproliferative activity against MCF-7 human breast cancer cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.