Learn More
Medchemexpress LLC Pinostrobin chalcone | 18956-15-5 | 5 MG
SDP
Catalog No. 5000421581
Encompass_Preferred
Pinostrobin chalcone is a potent natural cytotoxic compound with anti-inflammatory properties. It has shown cytotoxic effects against MDA-MB-231 and HT-29 colon cancer cell lines and exhibits anti-inflammatory activity by inhibiting LPS-induced IL-1beta mRNA expression in J774.A1 and RAW264.7 cells. This product is for research use only.
  • Potent natural cytotoxic compound
  • Exhibits cytotoxic effects against MDA-MB-231 and HT-29 colon cancer cell lines
  • Shows anti-inflammatory activity against mouse J774.A1 and RAW264.7 cells
  • Inhibits LPS-induced IL-1beta mRNA expression
  • Tested for antiproliferative activity against MCF-7 human breast cancer cells
Catalog No. 50-004-21581 Supplier Medchemexpress LLC Supplier No. HYN30575MG
Pinostrobin chalcone is a potent natural cytotoxic compound with anti-inflammatory properties. It has shown cytotoxic effects against MDA-MB-231 and HT-29 colon cancer cell lines and exhibits anti-inflammatory activity by inhibiting LPS-induced IL-1beta mRNA expression in J774.A1 and RAW264.7 cells. This product is for research use only.
  • Potent natural cytotoxic compound
  • Exhibits cytotoxic effects against MDA-MB-231 and HT-29 colon cancer cell lines
  • Shows anti-inflammatory activity against mouse J774.A1 and RAW264.7 cells
  • Inhibits LPS-induced IL-1beta mRNA expression
  • Tested for antiproliferative activity against MCF-7 human breast cancer cells