Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinostrobin chalcone | 18956-15-5 | 99.9% | 270.28 g/mol | C16H14O4 | 10 MG
SDP

Catalog No. 5000288980
Encompass_Preferred

Pinostrobin chalcone is a naturally occurring chalcone derivative supplied as a high-purity research standard for in vitro biological and analytical studies. It has reported cytotoxic activity against MDA-MB-231 and HT-29 cancer cell lines (IC50 = 20.42 ± 2.23 μg/mL and 22.51 ± 0.42 μg/mL). The compound is provided as a solid and should be stored sealed at 4°C, protected from moisture and light.

  • High purity (99.94%).
  • Molecular weight 270.28 g/mol.
  • Chemical formula C16H14O4.
  • CAS number 18956-15-5.
  • Suitable for in vitro cytotoxicity and analytical assays.
  • Provided in a 10 MG package for laboratory use.
  • Store at 4°C, sealed, away from moisture and light.

Catalog No. 50-002-88980 Supplier Medchemexpress LLC Supplier No. HYN305710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinostrobin chalcone is a naturally occurring chalcone derivative supplied as a high-purity research standard for in vitro biological and analytical studies. It has reported cytotoxic activity against MDA-MB-231 and HT-29 cancer cell lines (IC50 = 20.42 ± 2.23 μg/mL and 22.51 ± 0.42 μg/mL). The compound is provided as a solid and should be stored sealed at 4°C, protected from moisture and light.

  • High purity (99.94%).
  • Molecular weight 270.28 g/mol.
  • Chemical formula C16H14O4.
  • CAS number 18956-15-5.
  • Suitable for in vitro cytotoxicity and analytical assays.
  • Provided in a 10 MG package for laboratory use.
  • Store at 4°C, sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.