Learn More
Medchemexpress LLC Pinosylvin | 22139-77-1 | 5 MG
SDP
Catalog No. 5000421534
Encompass_Preferred
Pinosylvin is a pre-infectious stilbenoid toxin isolated from the heartwood of Pinus species, demonstrating anti-bacterial activities. This resveratrol analogue can induce cell apoptosis and autophagy in leukemia cells.
  • Pre-infectious stilbenoid toxin
  • Isolated from the heartwood of Pinus species
  • Exhibits anti-bacterial activities
  • Resveratrol analogue
  • Induces cell apoptosis and autophagy in leukemia cells
Catalog No. 50-004-21534 Supplier Medchemexpress LLC Supplier No. HYN23875MG
Pinosylvin is a pre-infectious stilbenoid toxin isolated from the heartwood of Pinus species, demonstrating anti-bacterial activities. This resveratrol analogue can induce cell apoptosis and autophagy in leukemia cells.
  • Pre-infectious stilbenoid toxin
  • Isolated from the heartwood of Pinus species
  • Exhibits anti-bacterial activities
  • Resveratrol analogue
  • Induces cell apoptosis and autophagy in leukemia cells