Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinosylvin monomethyl ether | 35302-70-6 | MFCD00210546 | 99.0% | 226.27 | C15H14O2 | 10 MG
SDP

Catalog No. 5000289159
Encompass_Preferred

Pinosylvin monomethyl ether is a small-molecule natural product used as an analytical and research standard. It has reported antibacterial and fungicidal activity and is supplied at high purity for laboratory studies.

  • Exhibits antibacterial and fungicidal activity.
  • High purity: 99.0%.
  • Molecular formula C15H14O2 and molecular weight 226.27.
  • CAS number 35302-70-6 for unambiguous identification.
  • Supplied as a 10 mg analytical sample; store at 4°C and protect from light.

Catalog No. 50-002-89159 Supplier Medchemexpress LLC Supplier No. HYN305610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinosylvin monomethyl ether is a small-molecule natural product used as an analytical and research standard. It has reported antibacterial and fungicidal activity and is supplied at high purity for laboratory studies.

  • Exhibits antibacterial and fungicidal activity.
  • High purity: 99.0%.
  • Molecular formula C15H14O2 and molecular weight 226.27.
  • CAS number 35302-70-6 for unambiguous identification.
  • Supplied as a 10 mg analytical sample; store at 4°C and protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.