Learn More
Zymo Research Corporation Pinpoint™ Slide RNA Isolation System I Kit (50 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. 501251657
Encompass_Preferred
The Pinpoint Slide RNA Isolation Systems I and II are innovative products for the isolation of RNA from any targeted area of fresh, paraffin-fixed, or paraffin-embedded tissues sectioned onto a glass slide. The system combines powerful Pinpoint tissue sampling methodology, a unique single-step RNA extraction/binding buffer, and Zymo-Spin column purification technology to yield high quality RNA. Unlike current UV-based methods, this product makes isolation of tissue RNA simple and quick. No expensive specialized equipment is needed. Eluted RNA is well suited for subsequent RNA analyses including RT-PCR.
Catalog No. 50-125-1657 Supplier Zymo Research Corporation Supplier No. R1003
The Pinpoint Slide RNA Isolation Systems I and II are innovative products for the isolation of RNA from any targeted area of fresh, paraffin-fixed, or paraffin-embedded tissues sectioned onto a glass slide. The system combines powerful Pinpoint tissue sampling methodology, a unique single-step RNA extraction/binding buffer, and Zymo-Spin column purification technology to yield high quality RNA. Unlike current UV-based methods, this product makes isolation of tissue RNA simple and quick. No expensive specialized equipment is needed. Eluted RNA is well suited for subsequent RNA analyses including RT-PCR.