Learn More
Instech PinPorts™, 22ga red, 10 sterile pouches of 5 (50 pcs)
SDP
Catalog No. 50890151
Encompass
The Instech PNP3F22R-50 is a specialized PinPort™ assembly designed for high-performance, aseptic access to catheters. This specific model features a 22-gauge recessed port, which is commonly used in laboratory research to allow for frequent, needle-free sampling or dosing while maintaining a closed system.
Features
  • Type: PinPort™ Catheter Access Port
  • Gauge: 22ga
  • Configuration: Recessed port (R)
  • Connection: 3Fr (F22) compatibility
  • Material: Medical-grade biocompatible plastics and stainless steel
  • Sterility: Pre-sterilized and individually pouched
  • Quantity: Box of 50 units
  • Application: Designed for exteriorized catheters to simplify aseptic access and improve animal welfare
Catalog No. 50-890-151 Supplier Instech Supplier No. PNP3F22R50
The Instech PNP3F22R-50 is a specialized PinPort™ assembly designed for high-performance, aseptic access to catheters. This specific model features a 22-gauge recessed port, which is commonly used in laboratory research to allow for frequent, needle-free sampling or dosing while maintaining a closed system.
Features
  • Type: PinPort™ Catheter Access Port
  • Gauge: 22ga
  • Configuration: Recessed port (R)
  • Connection: 3Fr (F22) compatibility
  • Material: Medical-grade biocompatible plastics and stainless steel
  • Sterility: Pre-sterilized and individually pouched
  • Quantity: Box of 50 units
  • Application: Designed for exteriorized catheters to simplify aseptic access and improve animal welfare