Learn More
Instech PinPorts™ for fast access to externalized catheters, 22ga white, 50 sterile pouches of 5 (250 pcs)
SDP
Catalog No. NC2816869
Encompass
PinPort™ for Catheters and Tubing
The PinPort™ provides fast, aseptic access to externalized rodent catheters or tubing for dosing or blood sampling. It can reduce study costs and improve compliance with the 3Rs. The system consists of a lightweight port and a mating injector which can access the port hundreds of times.
The PinPort™ is available with 22ga, 25ga or 28ga couplers to mate with 3Fr, 2Fr or PE10 catheters, respectively. The white or red colors can be used to distinguish vessels. Use only PNP3M injectors or PinPort™ connectors to access PinPorts™; any other type of needle will damage the port. All parts are provided sterilized by ethylene oxide.
Designed for short-term intermittent, manual access. It is not designed to be connected to a tether and swivel for remote or continuous access.
Features
  • Fast access
  • Closed system for improved patency
  • Needle free
  • Low cost
Catalog No. NC2816869 Supplier Instech Supplier No. PNP3F22
PinPort™ for Catheters and Tubing
The PinPort™ provides fast, aseptic access to externalized rodent catheters or tubing for dosing or blood sampling. It can reduce study costs and improve compliance with the 3Rs. The system consists of a lightweight port and a mating injector which can access the port hundreds of times.
The PinPort™ is available with 22ga, 25ga or 28ga couplers to mate with 3Fr, 2Fr or PE10 catheters, respectively. The white or red colors can be used to distinguish vessels. Use only PNP3M injectors or PinPort™ connectors to access PinPorts™; any other type of needle will damage the port. All parts are provided sterilized by ethylene oxide.
Designed for short-term intermittent, manual access. It is not designed to be connected to a tether and swivel for remote or continuous access.
Features
  • Fast access
  • Closed system for improved patency
  • Needle free
  • Low cost