Learn More
Instech PinPorts™, 25ga red, 10 sterile pouches of 5 (50 pcs)
SDP
Catalog No. 50890155
Encompass
The Instech PNP3F25R-50 is a sterile red 25ga PinPort™ set designed to provide fast, aseptic access to externalized laboratory animal catheters or tubing. Each port allows repeated dosing or sampling without needle use, supporting safer handling, improved catheter patency, and reduced contamination risk. The PNP3F25R-50 package includes ten sterile pouches, each containing five red 25ga PinPorts, making it suitable for multi-animal studies or high-throughput workflows.
Features
  • Port Type: PinPort™ access port
  • Gauge: 25ga
  • Color: Red
  • Packaging: 10 sterile pouches of 5 (50 total ports)
  • Use Case: Aseptic dosing and sampling of externalized catheters
  • Needle-Free Access: Supports safe, closed-system operation
  • Compatibility: Works with PNP3M injectors for manual access
Catalog No. 50-890-155 Supplier Instech Supplier No. PNP3F25R50
The Instech PNP3F25R-50 is a sterile red 25ga PinPort™ set designed to provide fast, aseptic access to externalized laboratory animal catheters or tubing. Each port allows repeated dosing or sampling without needle use, supporting safer handling, improved catheter patency, and reduced contamination risk. The PNP3F25R-50 package includes ten sterile pouches, each containing five red 25ga PinPorts, making it suitable for multi-animal studies or high-throughput workflows.
Features
  • Port Type: PinPort™ access port
  • Gauge: 25ga
  • Color: Red
  • Packaging: 10 sterile pouches of 5 (50 total ports)
  • Use Case: Aseptic dosing and sampling of externalized catheters
  • Needle-Free Access: Supports safe, closed-system operation
  • Compatibility: Works with PNP3M injectors for manual access