Learn More
Instech PinPorts™, 25ga red, with injector in sterile pouch (20 sets)
SDP
Catalog No. 50890160
Encompass
PinPort™ for Catheters and Tubing
Available with 22ga, 25ga, or 28ga couplers to pair with 3Fr, 2Fr, or PE10 catheters, respectively, the PinPort™ offers quick, aseptic access with clear vessel identification using white or red color options. Use only PNP3M injectors or PinPort™ connectors when accessing—any other needle will damage the port. All components arrive EO‑sterilized and ready to use.
Designed for short‑term, intermittent manual access, the PinPort™ is not intended for tethered or continuous setups. It must be positioned where the animal cannot reach it, which can be challenging in mice. For long‑term or swivel‑based applications, choose the Vascular Access Button™ for rats or mice.
PinPort, 25ga red, with injector in sterile pouch (20 sets)
Features
  • Fast access
  • Closed system for improved patency
  • Needle free
  • Low cost
Catalog No. 50-890-160 Supplier Instech Supplier No. PNP3MF25R
PinPort™ for Catheters and Tubing
Available with 22ga, 25ga, or 28ga couplers to pair with 3Fr, 2Fr, or PE10 catheters, respectively, the PinPort™ offers quick, aseptic access with clear vessel identification using white or red color options. Use only PNP3M injectors or PinPort™ connectors when accessing—any other needle will damage the port. All components arrive EO‑sterilized and ready to use.
Designed for short‑term, intermittent manual access, the PinPort™ is not intended for tethered or continuous setups. It must be positioned where the animal cannot reach it, which can be challenging in mice. For long‑term or swivel‑based applications, choose the Vascular Access Button™ for rats or mice.
PinPort, 25ga red, with injector in sterile pouch (20 sets)
Features
  • Fast access
  • Closed system for improved patency
  • Needle free
  • Low cost