Learn More
Instech Male PinPort™ connector, 25ga inlet, sterile
SDP
Catalog No. 50890162
Encompass
Use a PinPort™ connector to access a PinPort™ at the end of a segment of tubing. Applications include acute experiments where tethering is not necessary, or external tubing connections where the self-sealing septum in the PinPort™ is helpful.
The connectors are 12mm long and feature 25ga inputs. The tube that pierces the PinPort™ septum is 25ga. EO‑sterilized and ready to use.
Minimum order quantity 5 pcs.
For laboratory animal research use only.
Features
  • 25ga input
  • Low dead volume
  • Quick, clean connection
Catalog No. 50-890-162 Supplier Instech Supplier No. PNP3MC/25
Use a PinPort™ connector to access a PinPort™ at the end of a segment of tubing. Applications include acute experiments where tethering is not necessary, or external tubing connections where the self-sealing septum in the PinPort™ is helpful.
The connectors are 12mm long and feature 25ga inputs. The tube that pierces the PinPort™ septum is 25ga. EO‑sterilized and ready to use.
Minimum order quantity 5 pcs.
For laboratory animal research use only.
Features
  • 25ga input
  • Low dead volume
  • Quick, clean connection