Learn More
Instech PinPort extension set: PinPort connector, 12in (30cm) PU tubing, 25ga luer stub, sterile
SDP
Catalog No. NC1708858
Encompass
Use a PinPort™ connector to access a PinPort™ at the end of a segment of tubing. Applications include acute experiments where tethering is not necessary, or external tubing connections where the self-sealing septum in the PinPort™ is helpful.
The connectors are 12mm long and feature 22ga or 25ga inputs. The tube that pierces the PinPort™ septum is 25ga. The connectors can be built into extension sets with luers; see below for a standard 30cm set or contact us for custom lengths.
For laboratory animal research use only.
Features
  • 22ga or 25ga input
  • Low dead volume
  • Quick, clean connection
Catalog No. NC1708858 Supplier Instech Supplier No. PNP3MC/2512
Use a PinPort™ connector to access a PinPort™ at the end of a segment of tubing. Applications include acute experiments where tethering is not necessary, or external tubing connections where the self-sealing septum in the PinPort™ is helpful.
The connectors are 12mm long and feature 22ga or 25ga inputs. The tube that pierces the PinPort™ septum is 25ga. The connectors can be built into extension sets with luers; see below for a standard 30cm set or contact us for custom lengths.
For laboratory animal research use only.
Features
  • 22ga or 25ga input
  • Low dead volume
  • Quick, clean connection