Learn More
Instech PinPort™ injectors. For use with Instech PinPorts, mouse VAH harnesses and mouse VAB buttons. 50 sterile pouches of 5.
SDP
Catalog No. NC0637753
Encompass
The Instech PNP3M PinPort™ injector provides needle-free, aseptic access to externalized catheters through Instech PinPorts™, as well as mouse VAH harnesses and mouse VAB buttons. Its recessed blunted tip aligns with the PinPort™ septum for safe, repeatable dosing or sampling without causing damage. Supplied as 50 sterile pouches of 5, it connects via a standard luer for reliable manual access in rodent infusion and sampling workflows.
Features
  • Injector for PinPorts™, mouse VAH and VAB
  • Recessed, needle-free tip
  • Standard luer connection
  • Protects PinPort™ septa
  • 50 sterile pouches of 5
  • EtO sterilized
Catalog No. NC0637753 Supplier Instech Supplier No. PNP3M
The Instech PNP3M PinPort™ injector provides needle-free, aseptic access to externalized catheters through Instech PinPorts™, as well as mouse VAH harnesses and mouse VAB buttons. Its recessed blunted tip aligns with the PinPort™ septum for safe, repeatable dosing or sampling without causing damage. Supplied as 50 sterile pouches of 5, it connects via a standard luer for reliable manual access in rodent infusion and sampling workflows.
Features
  • Injector for PinPorts™, mouse VAH and VAB
  • Recessed, needle-free tip
  • Standard luer connection
  • Protects PinPort™ septa
  • 50 sterile pouches of 5
  • EtO sterilized