Learn More
Instech PinPort™ injectors. For use with Instech PinPorts, mouse VAH harnesses and mouse VAB buttons. 10 sterile pouches of 5.
SDP
Catalog No. NC0629377
Encompass
The Instech PNP3M-50 PinPort™ injector provides fast, needle-free access to externalized catheters via Instech PinPorts™, and is also used with mouse VAH harnesses and mouse VAB buttons. Its recessed blunted tip aligns with the PinPort™ septum for safe, repeatable dosing or sampling without damage. Supplied as 10 sterile pouches of 5, it connects through a standard luer for quick manual access in rodent infusion workflows.
Features
  • Injector for PinPorts™, mouse VAH and VAB
  • Recessed, needle-free tip
  • Standard luer connection
  • Protects PinPort™ septa
  • 10 sterile pouches of 5
  • EtO sterilized
Catalog No. NC0629377 Supplier Instech Supplier No. PNP3M50
The Instech PNP3M-50 PinPort™ injector provides fast, needle-free access to externalized catheters via Instech PinPorts™, and is also used with mouse VAH harnesses and mouse VAB buttons. Its recessed blunted tip aligns with the PinPort™ septum for safe, repeatable dosing or sampling without damage. Supplied as 10 sterile pouches of 5, it connects through a standard luer for quick manual access in rodent infusion workflows.
Features
  • Injector for PinPorts™, mouse VAH and VAB
  • Recessed, needle-free tip
  • Standard luer connection
  • Protects PinPort™ septa
  • 10 sterile pouches of 5
  • EtO sterilized