Learn More
Instech PinPort™ for fast access to externalized catheters. 25ga connector, white. 10 sterile pouches of 5.
SDP
Catalog No. NC1849965
Encompass
The Instech PinPort PNP3F25-50 is a mid-sized access port designed for aseptic, needle-free sampling or dosing in laboratory research. This specific part number represents a bulk package of 50 units, organized into 10 sterile pouches of 5 ports each. These ports are constructed to provide a closed system that protects catheter patency and conforms to 3Rs animal welfare standards by eliminating the need for sharp needles during routine access.
Features
  • Part Number: PNP3F25-50
  • Coupler Size: 25ga
  • Compatibility: Mates with 2Fr catheters
  • Color: White
  • Sterilization: Ethylene Oxide (EtO)
  • Quantity: 50 pieces (10 pouches of 5)
  • Design: Low-profile for intermittent manual access
  • Maintenance: Requires specialized PNP3M injectors to maintain seal integrity
Catalog No. NC1849965 Supplier Instech Supplier No. PNP3F2550
The Instech PinPort PNP3F25-50 is a mid-sized access port designed for aseptic, needle-free sampling or dosing in laboratory research. This specific part number represents a bulk package of 50 units, organized into 10 sterile pouches of 5 ports each. These ports are constructed to provide a closed system that protects catheter patency and conforms to 3Rs animal welfare standards by eliminating the need for sharp needles during routine access.
Features
  • Part Number: PNP3F25-50
  • Coupler Size: 25ga
  • Compatibility: Mates with 2Fr catheters
  • Color: White
  • Sterilization: Ethylene Oxide (EtO)
  • Quantity: 50 pieces (10 pouches of 5)
  • Design: Low-profile for intermittent manual access
  • Maintenance: Requires specialized PNP3M injectors to maintain seal integrity