Learn More
StatLab Pinworm Collector Kit - Direct-to-Slide
SDP
Catalog No. 22130481
Change view
Click to view available options
Quantity:
25/Pk.
Catalog No. Quantity
22-130-481 25/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-130-481 Supplier StatLab Supplier No. 582
Add to Cart
Add to Cart
This item is not returnable. View return policy
See ova more precisely with this clear and thinner polystyrene paddle
  • Each Pinworm Collector has a paddle with one 'sticky side' to allow ova to stick to the adhesive
  • View collection paddle directly on your microscope by simply snapping off top of vial and applying to slide
  • Paddle then becomes the coverslip
  • 25 vials/box
Specifications
Product Type Pinworm Paddle
Format Paddle
Sterility Non-sterile
Quantity 25/Pk.