Learn More
Aviva Systems Biology Corp PINX1 Antibody - middle region (ARP57058_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57058_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PINX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PINX1. Alternate names: FLJ20565, LPTL, LPTS, MGC8850ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-261