Learn More
Cayman Chemical pIodoClonIdInhydrochLRI 10mg
SDP
Catalog No. 502966238
Encompass_Preferred
A partial agonist of the α2-adrenergic receptor, minimally inhibiting adenylate cyclase through this Gi protein-coupled receptor although it binds avidly (Ki = 1.0 nM); potentiates platelet aggregation by ADP (EC50 = 1.5 µM) and inhibits epinephrine-induced aggregation (IC50 = 5.1 µM)
Catalog No. 50-296-6238 Supplier Cayman Chemical Supplier No. 1717710 MG
A partial agonist of the α2-adrenergic receptor, minimally inhibiting adenylate cyclase through this Gi protein-coupled receptor although it binds avidly (Ki = 1.0 nM); potentiates platelet aggregation by ADP (EC50 = 1.5 µM) and inhibits epinephrine-induced aggregation (IC50 = 5.1 µM)