Learn More
Cayman Chemical Pioglitazone Ketone, Purity: ≥98%, CAS: 146062-45-5, 5 mg
SDP
Catalog No. 502939458
Encompass_Preferred
An active metabolite of pioglitazone; formed primarly through CYP2C8-mediated metabolism of pioglitazone
Catalog No. 50-293-9458 Supplier Cayman Chemical Supplier No. 280445 MG
An active metabolite of pioglitazone; formed primarly through CYP2C8-mediated metabolism of pioglitazone