Learn More
TARGETMOL CHEMICALS INC PIOGLITAZONE 100MG
SDP
Catalog No. 502581791
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pioglitazone (U 72107) is an insulin sensitizing agent and thiazolidinedione that is indicated for the treatment of type 2 diabetes. Pioglitazone has been linked to rare instances of acute liver injury. purity: 99%
Catalog No. 50-258-1791 Supplier TARGETMOL CHEMICALS INC Supplier No. T0214100MG
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pioglitazone (U 72107) is an insulin sensitizing agent and thiazolidinedione that is indicated for the treatment of type 2 diabetes. Pioglitazone has been linked to rare instances of acute liver injury. purity: 99%