Learn More
Apexbio Technology LLC Pioglitazone(Synonyms: Actos, Pioglitazone Hydrochloride, AD-4833, U-72107, Glustin, Zactos, Piozone), 10mM (in 1mL DMSO), CAS: 111025-46-8.
SDP
Catalog No. 5000060153
Encompass_Preferred
Pioglitazone (CAS 111025-46-8) is a small-molecule agonist selectively targeting peroxisome proliferator-activated receptor gamma (PPAR ) By activating this nuclear receptor pioglitazone influences gene expression pathways involved primarily in glucose and lipid metabolism insulin sensitivity and adipocyte differentiation It is widely employed in biomedical research focusing on metabolic disorders including type 2 diabetes mellitus insulin resistance mechanisms and associated inflammatory processes
Catalog No. 50-000-60153 Supplier Apexbio Technology LLC Supplier No. B21175.1
Pioglitazone (CAS 111025-46-8) is a small-molecule agonist selectively targeting peroxisome proliferator-activated receptor gamma (PPAR ) By activating this nuclear receptor pioglitazone influences gene expression pathways involved primarily in glucose and lipid metabolism insulin sensitivity and adipocyte differentiation It is widely employed in biomedical research focusing on metabolic disorders including type 2 diabetes mellitus insulin resistance mechanisms and associated inflammatory processes