Learn More
Enzo Life Sciences Pioglitazone (1mg). CAS: 111025-46-8
SDP
Supplier:  Enzo Life Sciences ALX270367M001
Encompass_Preferred
PPARγ activator. Pioglitazone selectively activates PPARγ-1. It is about one tenth as potent as rosiglitazone (EC50~500nM for human and mouse PPARγ). In a transactivation assay using COS-1 cells transfected with full length human PPARα and RXRα, pioglitazone and rosiglitazone exhibit low level activation of PPARα at 1µM and 5.4- and 4.2-fold activation, respectively, at a concentration of 10µM. Formula: C19H20N2O3S. MW: 356.4. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (2.5mg/ml) or dimethyl formamide (2.5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8512