Learn More
Selleck Chemical LLC Pioglitazone 10mM/1mL 111025-46-8
SDP
Supplier:  Selleck Chemical LLC S259010MM/1ML
Encompass_Preferred
Pioglitazone is a selective peroxisome proliferator-activated receptor-gamma (PPAR?) agonist, used to treat diabetes; A weak activator for full-length hPPAR?, but not full-length hPPAR?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2599