Learn More
AdipoGen Pioglitazone
SDP
Supplier:  AdipoGen AGCR10067M025
Encompass_Preferred
Chemical. CAS 111025-46-8. Formula C19H20N2O3S. MW 356.4. Thiazolidinedione TZD reference compound. Selective PPARgamma agonist. Selectively activates PPARgamma-1. Anti-diabetic , decreasing insulin resistance in adipose tissue. Is about one tenth as potent as rosiglitazone. Lipid metabolism modulator. Estrogen synthesis inhibitor. NF-kappaB inhibitor. Neuroprotective.
Catalog No. 50-256-2371