Learn More
AdipoGen Pioglitazone
SDP
Catalog No. 502562372
Encompass_Preferred
Chemical. CAS 111025-46-8. Formula C19H20N2O3S. MW 356.4. Thiazolidinedione TZD reference compound. Selective PPARgamma agonist. Selectively activates PPARgamma-1. Anti-diabetic , decreasing insulin resistance in adipose tissue. Is about one tenth as potent as rosiglitazone. Lipid metabolism modulator. Estrogen synthesis inhibitor. NF-kappaB inhibitor. Neuroprotective.
Catalog No. 50-256-2372 Supplier AdipoGen Supplier No. AGCR10067M005
Chemical. CAS 111025-46-8. Formula C19H20N2O3S. MW 356.4. Thiazolidinedione TZD reference compound. Selective PPARgamma agonist. Selectively activates PPARgamma-1. Anti-diabetic , decreasing insulin resistance in adipose tissue. Is about one tenth as potent as rosiglitazone. Lipid metabolism modulator. Estrogen synthesis inhibitor. NF-kappaB inhibitor. Neuroprotective.