Learn More
Selleck Chemical LLC Pioglitazone S2590-200mg
SDP
Supplier:  Selleck Chemical LLC S2590200MG
Encompass_Preferred
Pioglitazone is a selective peroxisome proliferator-activated receptor-gamma (PPAR ) agonist used to treat diabetes A weak activator for full-length hPPAR but not full-length hPPAR
Catalog No. 50-311-0228