Learn More
Apexbio Technology LLC Pioglitazone HCl 112529-15-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062539
Encompass_Preferred
Pioglitazone HCl is a potent and highly selective agonist of the peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor involved in glucose and lipid metabolism regulation Acting via PPAR activation pioglitazone increases insulin sensitivity in hepatic adipose and peripheral tissues enhancing insulin-mediated glucose uptake and utilization Pioglitazone requires the presence of insulin to exert its biological activity through receptor activation limiting its application in scenarios characterized by substantial insulin deficiency Clinically pioglitazone HCl has been widely applied in the management and investigation of hyperglycemia associated with type 2 diabetes mellitus (T2DM) Additionally pioglitazone demonstrates effects on lipid regulation notably reducing triglyceride levels and increasing high-density lipoprotein (HDL) cholesterol making it useful in metabolic research
Catalog No. 50-000-62539 Supplier Apexbio Technology LLC Supplier No. A43245.1
Pioglitazone HCl is a potent and highly selective agonist of the peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor involved in glucose and lipid metabolism regulation Acting via PPAR activation pioglitazone increases insulin sensitivity in hepatic adipose and peripheral tissues enhancing insulin-mediated glucose uptake and utilization Pioglitazone requires the presence of insulin to exert its biological activity through receptor activation limiting its application in scenarios characterized by substantial insulin deficiency Clinically pioglitazone HCl has been widely applied in the management and investigation of hyperglycemia associated with type 2 diabetes mellitus (T2DM) Additionally pioglitazone demonstrates effects on lipid regulation notably reducing triglyceride levels and increasing high-density lipoprotein (HDL) cholesterol making it useful in metabolic research